Anti-CLEC4N (alpha Isoform) Rat Monoclonal Antibody [clone: 217611]

Supplier: R & D Systems
MAB1525
220019-149EA 763.57 USD
220019-149
Anti-CLEC4N (alpha Isoform) Rat Monoclonal Antibody [clone: 217611]
Antibodies
Type: Primary
Antigen: CLEC4N (alpha Isoform)
Clonality: Monoclonal
Clone: 217611
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR