Anti-PODXL2 Mouse Monoclonal Antibody [clone: 211805]

Supplier: R & D Systems
MAB15241
220019-148EA 763.57 USD
220019-148
Anti-PODXL2 Mouse Monoclonal Antibody [clone: 211805]
Antibodies
Type: Primary
Antigen: PODXL2
Clonality: Monoclonal
Clone: 211805
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR