Anti-PODXL2 Mouse Monoclonal Antibody [clone: 211816]

Supplier: R & D Systems
MAB1524
220019-147EA 594.37 USD
220019-147
Anti-PODXL2 Mouse Monoclonal Antibody [clone: 211816]
Antibodies
Type: Primary
Antigen: PODXL2
Clonality: Monoclonal
Clone: 211816
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR