Anti-ULBP3 Mouse Monoclonal Antibody [clone: 166514]

Supplier: R & D Systems
MAB15171
220019-144EA 763.57 USD
220019-144
Anti-ULBP3 Mouse Monoclonal Antibody [clone: 166514]
Antibodies
Type: Primary
Antigen: ULBP3
Clonality: Monoclonal
Clone: 166514
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR