Anti-2010005H15RIK Rat Monoclonal Antibody [clone: 23735]

Supplier: R & D Systems
MAB1516
220019-142EA 763.57 USD
220019-142
Anti-2010005H15RIK Rat Monoclonal Antibody [clone: 23735]
Antibodies
Type: Primary
Antigen: 2010005H15RIK
Clonality: Monoclonal
Clone: 23735
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR