Anti-ACE Rat Monoclonal Antibody [clone: 2325]

Supplier: R & D Systems
MAB15132
220019-141EA 763.57 USD
220019-141
Anti-ACE Rat Monoclonal Antibody [clone: 2325]
Antibodies
Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone: 2325
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR