Anti-ACE Rat Monoclonal Antibody [clone: 23214]

Supplier: R & D Systems
MAB15131
220019-140EA 561.11 USD
220019-140
Anti-ACE Rat Monoclonal Antibody [clone: 23214]
Antibodies
Type: Primary
Antigen: ACE
Clonality: Monoclonal
Clone: 23214
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR