Anti-CD8A Mouse Monoclonal Antibody [clone: 37006]

Supplier: R & D Systems
MAB1509
220019-137EA 763.57 USD
220019-137
Anti-CD8A Mouse Monoclonal Antibody [clone: 37006]
Antibodies
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 37006
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR