Anti-DKKL1 Rat Monoclonal Antibody [clone: 192425]

Supplier: R & D Systems
MAB1508
220019-136EA 763.57 USD
220019-136
Anti-DKKL1 Rat Monoclonal Antibody [clone: 192425]
Antibodies
Type: Primary
Antigen: DKKL1
Clonality: Monoclonal
Clone: 192425
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR