Anti-TNF Mouse Monoclonal Antibody [clone: 236812]

Supplier: R & D Systems
MAB1507
220019-135EA 772.25 USD
220019-135
Anti-TNF Mouse Monoclonal Antibody [clone: 236812]
Antibodies
Type: Primary
Antigen: TNF
Clonality: Monoclonal
Clone: 236812
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR