Anti-IL17D Mouse Monoclonal Antibody [clone: 246018]

Supplier: R & D Systems
MAB15041
220019-134EA 594.37 USD
220019-134
Anti-IL17D Mouse Monoclonal Antibody [clone: 246018]
Antibodies
Type: Primary
Antigen: IL17D
Clonality: Monoclonal
Clone: 246018
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR