Anti-FURIN Mouse Monoclonal Antibody [clone: 222728]

Supplier: R & D Systems
MAB15031
220019-131EA 763.57 USD
220019-131
Anti-FURIN Mouse Monoclonal Antibody [clone: 222728]
Antibodies
Type: Primary
Antigen Symbol: FURIN
Antigen Name: furin (paired basic amino acid cleaving enzyme)
Antigen Synonyms: FUR, PACE, PCSK3, SPC1
Clonality: Monoclonal
Clone: 222728
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR