Anti-CCR2 Mouse Monoclonal Antibody [clone: 48607]

Supplier: R & D Systems
MAB150-100
102885-132EA 647.88 USD
102885-132
Anti-CCR2 Mouse Monoclonal Antibody [clone: 48607]
Antibodies
Type: Primary
Antigen: CCR2
Clonality: Monoclonal
Clone: 48607
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR