Anti-CTS7 Rat Monoclonal Antibody [clone: 228905]

Supplier: R & D Systems
MAB1499
220019-128EA 763.57 USD
220019-128
Anti-CTS7 Rat Monoclonal Antibody [clone: 228905]
Antibodies
Type: Primary
Antigen: CTS7
Clonality: Monoclonal
Clone: 228905
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR