Anti-EPHA7 Rat Monoclonal Antibody [clone: 88712]

Supplier: R & D Systems
MAB1495
220019-127EA 763.57 USD
220019-127
Anti-EPHA7 Rat Monoclonal Antibody [clone: 88712]
Antibodies
Type: Primary
Antigen: EPHA7
Clonality: Monoclonal
Clone: 88712
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR