Anti-STAT1 Mouse Monoclonal Antibody [clone: 655210]

Supplier: R & D Systems
MAB14901
102885-130EA 594.37 USD
102885-130
Anti-STAT1 Mouse Monoclonal Antibody [clone: 655210]
Antibodies
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 655210
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR