Anti-STAT1 Rat Monoclonal Antibody [clone: 246523]

Supplier: R & D Systems
MAB1490
220019-124EA 594.37 USD
220019-124
Anti-STAT1 Rat Monoclonal Antibody [clone: 246523]
Antibodies
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: 246523
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR