Anti-PSEN1 Mouse Monoclonal Antibody [clone: 121121]

Supplier: R & D Systems
MAB149
220019-123EA 763.57 USD
220019-123
Anti-PSEN1 Mouse Monoclonal Antibody [clone: 121121]
Antibodies
Type: Primary
Antigen: PSEN1
Clonality: Monoclonal
Clone: 121121
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR