Anti-TLR3 Mouse Monoclonal Antibody [clone: 512505]

Supplier: R & D Systems
MAB1487
220019-122EA 594.37 USD
220019-122
Anti-TLR3 Mouse Monoclonal Antibody [clone: 512505]
Antibodies
Type: Primary
Antigen: TLR3
Clonality: Monoclonal
Clone: 512505
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR