Anti-BCAM Mouse Monoclonal Antibody [clone: 87207]

Supplier: R & D Systems
MAB1481
220019-121EA 763.57 USD
220019-121
Anti-BCAM Mouse Monoclonal Antibody [clone: 87207]
Antibodies
Type: Primary
Antigen: BCAM
Clonality: Monoclonal
Clone: 87207
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR