Anti-BCAM Mouse Monoclonal Antibody [clone: 87202]

Supplier: R & D Systems
MAB148
220019-120EA 763.57 USD
220019-120
Anti-BCAM Mouse Monoclonal Antibody [clone: 87202]
Antibodies
Type: Primary
Antigen: BCAM
Clonality: Monoclonal
Clone: 87202
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR