Anti-IL27RA Mouse Monoclonal Antibody [clone: 191106]

Supplier: R & D Systems
MAB1479
220019-119EA 779.48 USD
220019-119
Anti-IL27RA Mouse Monoclonal Antibody [clone: 191106]
Antibodies
Type: Primary
Antigen: IL27RA
Clonality: Monoclonal
Clone: 191106
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR