Anti-TLR4 Mouse Monoclonal Antibody [clone: 610017]

Supplier: R & D Systems
MAB14783
220019-118EA 577.02 USD
220019-118
Anti-TLR4 Mouse Monoclonal Antibody [clone: 610017]
Antibodies
Type: Primary
Antigen: TLR4
Clonality: Monoclonal
Clone: 610017
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR