Anti-ACVR1B Rat Monoclonal Antibody [clone: 2734]

Supplier: R & D Systems
MAB1477
220019-115EA 763.57 USD
220019-115
Anti-ACVR1B Rat Monoclonal Antibody [clone: 2734]
Antibodies
Type: Primary
Antigen: ACVR1B
Clonality: Monoclonal
Clone: 2734
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR