Anti-TLR1 Rat Monoclonal Antibody [clone: 285923]

Supplier: R & D Systems
MAB1475
220019-114EA 577.02 USD
220019-114
Anti-TLR1 Rat Monoclonal Antibody [clone: 285923]
Antibodies
Type: Primary
Antigen: TLR1
Clonality: Monoclonal
Clone: 285923
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR