Anti-IL15RA Mouse Monoclonal Antibody [clone: 151307]

Supplier: R & D Systems
MAB147
220019-109EA 779.48 USD
220019-109
Anti-IL15RA Mouse Monoclonal Antibody [clone: 151307]
Antibodies
Type: Primary
Antigen: IL15RA
Clonality: Monoclonal
Clone: 151307
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR