Anti-AKT3 Mouse Monoclonal Antibody [clone: 199822]

Supplier: R & D Systems
MAB14631
220019-108EA 566.89 USD
220019-108
Anti-AKT3 Mouse Monoclonal Antibody [clone: 199822]
Antibodies
Type: Primary
Antigen: AKT3
Clonality: Monoclonal
Clone: 199822
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR