Anti-IL13RA1 Mouse Monoclonal Antibody [clone: 419718]

Supplier: R & D Systems
MAB1462
220019-106EA 600.15 USD
220019-106
Anti-IL13RA1 Mouse Monoclonal Antibody [clone: 419718]
Antibodies
Type: Primary
Antigen: IL13RA1
Clonality: Monoclonal
Clone: 419718
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR