Anti-CD32/CD16 Rat Monoclonal Antibody [clone: 190909]

Supplier: R & D Systems
MAB1460
220019-104EA 763.57 USD
220019-104
Anti-CD32/CD16 Rat Monoclonal Antibody [clone: 190909]
Antibodies
Type: Primary
Antigen: CD32/CD16
Clonality: Monoclonal
Clone: 190909
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR