Anti-IL13RA1 Mouse Monoclonal Antibody [clone: 116730]

Supplier: R & D Systems
MAB146
220019-103EA 772.25 USD
220019-103
Anti-IL13RA1 Mouse Monoclonal Antibody [clone: 116730]
Antibodies
Type: Primary
Antigen: IL13RA1
Clonality: Monoclonal
Clone: 116730
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR