Anti-HTRA2 Mouse Monoclonal Antibody [clone: 229926]

Supplier: R & D Systems
MAB1458
220019-102EA 577.02 USD
220019-102
Anti-HTRA2 Mouse Monoclonal Antibody [clone: 229926]
Antibodies
Type: Primary
Antigen: HTRA2
Clonality: Monoclonal
Clone: 229926
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR