Anti-ECE3 Rat Monoclonal Antibody [clone: 214711]

Supplier: R & D Systems
MAB1454
220019-100EA 763.57 USD
220019-100
Anti-ECE3 Rat Monoclonal Antibody [clone: 214711]
Antibodies
Type: Primary
Antigen: ECE3
Clonality: Monoclonal
Clone: 214711
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR