Anti-ALPL Mouse Monoclonal Antibody [clone: B4-78]

Supplier: R & D Systems
MAB1448
220019-098EA 647.88 USD
220019-098
Anti-ALPL Mouse Monoclonal Antibody [clone: B4-78]
Antibodies
Type: Primary
Antigen: ALPL
Clonality: Monoclonal
Clone: B4-78
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR