Anti-PDGFC Rat Monoclonal Antibody [clone: 184129]

Supplier: R & D Systems
MAB1447
220019-097EA 763.57 USD
220019-097
Anti-PDGFC Rat Monoclonal Antibody [clone: 184129]
Antibodies
Type: Primary
Antigen: PDGFC
Clonality: Monoclonal
Clone: 184129
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR