Anti-MIS/AMH Mouse Monoclonal Antibody [clone: 187406]

Supplier: R & D Systems
MAB1446
220019-096EA 763.57 USD
220019-096
Anti-MIS/AMH Mouse Monoclonal Antibody [clone: 187406]
Antibodies
Type: Primary
Antigen: MIS/AMH
Clonality: Monoclonal
Clone: 187406
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR