Anti-PRL Rat Monoclonal Antibody [clone: 207518]

Supplier: R & D Systems
MAB1445
220019-095EA 763.57 USD
220019-095
Anti-PRL Rat Monoclonal Antibody [clone: 207518]
Antibodies
Type: Primary
Antigen: PRL
Clonality: Monoclonal
Clone: 207518
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR