Anti-ANGPTL2 Rat Monoclonal Antibody [clone: 21]

Supplier: R & D Systems
MAB1444
220019-094EA 763.57 USD
220019-094
Anti-ANGPTL2 Rat Monoclonal Antibody [clone: 21]
Antibodies
Type: Primary
Antigen: ANGPTL2
Clonality: Monoclonal
Clone: 21
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR