Anti-SLC2A2 Rat Monoclonal Antibody [clone: 205115]

Supplier: R & D Systems
MAB1440
220019-092EA 623.29 USD
220019-092
Anti-SLC2A2 Rat Monoclonal Antibody [clone: 205115]
Antibodies
Type: Primary
Antigen: SLC2A2
Clonality: Monoclonal
Clone: 205115
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR