Anti-GFRA4 Mouse Monoclonal Antibody [clone: 215725]

Supplier: R & D Systems
MAB1439
220019-090EA 763.57 USD
220019-090
Anti-GFRA4 Mouse Monoclonal Antibody [clone: 215725]
Antibodies
Type: Primary
Antigen Symbol: GFRA4
Antigen Name: GDNF family receptor alpha 4
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 215725
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR