Anti-PARK2 Mouse Monoclonal Antibody [clone: 328122]

Supplier: R & D Systems
MAB14381
220019-089EA 561.11 USD
220019-089
Anti-PARK2 Mouse Monoclonal Antibody [clone: 328122]
Antibodies
Type: Primary
Antigen: PARK2
Clonality: Monoclonal
Clone: 328122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR