Anti-PARK2 Mouse Monoclonal Antibody [clone: 323302]

Supplier: R & D Systems
MAB1438
220019-088EA 561.11 USD
220019-088
Anti-PARK2 Mouse Monoclonal Antibody [clone: 323302]
Antibodies
Type: Primary
Antigen: PARK2
Clonality: Monoclonal
Clone: 323302
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR