Anti-IGFALS Rat Monoclonal Antibody [clone: 199410]

Supplier: R & D Systems
MAB1436
220019-087EA 763.57 USD
220019-087
Anti-IGFALS Rat Monoclonal Antibody [clone: 199410]
Antibodies
Type: Primary
Antigen: IGFALS
Clonality: Monoclonal
Clone: 199410
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR