Anti-SSEA3 Rat Monoclonal Antibody [clone: MC-631]

Supplier: R & D Systems
MAB1434
220019-085EA 594.37 USD
220019-085
Anti-SSEA3 Rat Monoclonal Antibody [clone: MC-631]
Antibodies
Type: Primary
Antigen: SSEA3
Clonality: Monoclonal
Clone: MC-631
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgM
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR