Anti-SPP1 Mouse Monoclonal Antibody [clone: 190312]

Supplier: R & D Systems
MAB1433
220019-082EA 600.15 USD
220019-082
Anti-SPP1 Mouse Monoclonal Antibody [clone: 190312]
Antibodies
Type: Primary
Antigen: SPP1
Clonality: Monoclonal
Clone: 190312
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR