Anti-CCR8 Rat Monoclonal Antibody [clone: 191704]

Supplier: R & D Systems
MAB1429-100
102885-270EA 647.88 USD
102885-270
Anti-CCR8 Rat Monoclonal Antibody [clone: 191704]
Antibodies
Type: Primary
Antigen: CCR8
Clonality: Monoclonal
Clone: 191704
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR