Anti-EPHB6 Rat Monoclonal Antibody [clone: 92224]

Supplier: R & D Systems
MAB1428
220019-077EA 763.57 USD
220019-077
Anti-EPHB6 Rat Monoclonal Antibody [clone: 92224]
Antibodies
Type: Primary
Antigen: EPHB6
Clonality: Monoclonal
Clone: 92224
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR