Anti-EREG Mouse Monoclonal Antibody [clone: 183629]

Supplier: R & D Systems
MAB14251
220019-074EA 577.02 USD
220019-074
Anti-EREG Mouse Monoclonal Antibody [clone: 183629]
Antibodies
Type: Primary
Antigen: EREG
Clonality: Monoclonal
Clone: 183629
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR