Anti-ACTA2 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: R & D Systems
MAB1420
220019-071EA 647.88 USD
220019-071
Anti-ACTA2 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
Type: Primary
Antigen: ACTA2
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR