Anti-FAS Mouse Monoclonal Antibody [clone: DX2]

Supplier: R & D Systems
MAB142
220019-070EA 763.57 USD
220019-070
Anti-FAS Mouse Monoclonal Antibody [clone: DX2]
Antibodies
Type: Primary
Antigen Symbol: FAS
Antigen Name: Fas cell surface death receptor
Antigen Synonyms: ALPS1A, APO-1, APT1, CD95, FAS1, FASTM, TNFRSF6
Clonality: Monoclonal
Clone: DX2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR