Anti-BGLAP Mouse Monoclonal Antibody [clone: 190125]

Supplier: R & D Systems
MAB1419
220019-069EA 647.88 USD
220019-069
Anti-BGLAP Mouse Monoclonal Antibody [clone: 190125]
Antibodies
Type: Primary
Antigen: BGLAP
Clonality: Monoclonal
Clone: 190125
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR